(S)-2,2'-Dimethoxy-1,1'-binaphthyl ≥98.0% (by GC)
TCIAD2821-1G
: TCI
:
- Pk:1 g
- Seq:3047
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:228.0 to 232.0 deg-C
- Specific rotation [a]20/D:-70.0 to -75.0 deg (C=1.2, THF)
:
:314.38 g/mol
:75640-87-8
:C₂₂H₁₈O₂
:MFCD00091146