(R)-(+)-2-Diphenylphosphino-2'-methoxy-1,1'-binaphthyl ≥98.0% (by GC)
: TCI
:145964-33-6
:
:C₃₃H₂₅OP
:MFCD00269686
:468.54 g/mol
- Appearance:White to Light yellow powder to crystal
- Purity (GC):min. 98.0 %
- Specific rotation [a]20/D:+91.0 to +98.0 deg (C=0.3, CHCl3)
- Solubility in Toluene:almost transparency
Frequently Bought Together