Order Entry
Italy
ContactUsLinkComponent
4-Diphenylphosphinomethyl polystyrene resin cross-linked with 2% DVB (200-400 mesh)
4-Diphenylphosphinomethyl polystyrene resin cross-linked with 2% DVB (200-400 mesh)
Catalog # TCIAD2766-5G
Supplier:  TCI
CAS Number:  
undefined
4-Diphenylphosphinomethyl polystyrene resin cross-linked with 2% DVB (200-400 mesh)
Catalog # TCIAD2766-5G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
MDL Number:MFCD03701782