5,5'-Dibromo-2,2'-bithiophene ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:4805-22-5
:
Formula:C₈H₄Br₂S₂
MDL Number:MFCD00219110
:324.05 g/mol
Specification Test Results
- Purity (GC):min. 98.0 %
- Melting point:144.0 to 148.0 deg-C
Specification Test Results
Frequently Bought Together