(R)-2,2'-Dimethoxy-1,1'-binaphthyl ≥98.0% (by GC)
Catalog # TCIAD2747-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:230.0 to 234.0 deg-C
- Specific rotation [a]20/D:+70.0 to +75.0 deg (C=1.2, THF)
Specification Test Results
About this item
:
:314.38 g/mol
CAS Number:35294-28-1
Formula:C₂₂H₁₈O₂
MDL Number:MFCD00091146