4-(Di-p-tolylamino)benzaldehyde ≥98.0% (by HPLC)
Supplier: TCI
Certificates
About this item
CAS Number:42906-19-4
:
Formula:C₂₁H₁₉NO
MDL Number:MFCD03093257
:301.39 g/mol
EINECS:255-996-1
Specification Test Results
- Appearance:Light yellow to Amber to Dark green powder to crystal
- Purity (HPLC):min. 98.0 area%
- Melting point:107.0 to 111.0 deg-C
- Solubility in hot Acetonitrile:almost transparency
Specification Test Results
Frequently Bought Together