2,7-Dimethoxy-1,4,5,8-tetrahydronaphthalene ≥97.0% (by GC)
Catalog # TCIAD2732-5G
Supplier: TCI
CAS Number:
Specifications
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 97.0 %
- Melting point:64.0 to 68.0 deg-C
Specification Test Results
About this item
:
:192.26 g/mol
Boiling Pt:100°C
CAS Number:1614-82-0
Formula:C₁₂H₁₆O₂
MDL Number:MFCD00269695