
4,4'-Dihydroxytetraphenylmethane ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:1844-01-5
:
Formula:C₂₅H₂₀O₂
MDL Number:MFCD00045785
:352.43 g/mol
:GHS07
Specification Test Results
- Appearance:White to Orange to Green powder to crystal
- Purity (GC):min. 98.0 %
- Solubility in Methanol:almost transparency
Specification Test Results
Frequently Bought Together