Order Entry
Italy
ContactUsLinkComponent
4,5-Dimethylfurfural ≥97.0% (by GC)
4,5-Dimethylfurfural ≥97.0% (by GC)
Catalog # TCIAD2639-1G
Supplier:  TCI
CAS Number:  
4,5-Dimethylfurfural ≥97.0% (by GC)
Catalog # TCIAD2639-1G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Appearance:
    White to Almost white clear liquid
  • Purity (GC):
    min. 97.0 %

Specification Test Results

About this item

 :
 :124.14 g/mol
Boiling Pt:100°C
Flash Pt:66°C
CAS Number:52480-43-0
Formula:C₇H₈O₂
MDL Number:MFCD00085030