1,2:3,5-Di-O-isopropylidene-ɑ-D-xylofuranose ≥98.0% (by GC)
Catalog # TCIAD2616-5G
Supplier: TCI
CAS Number:
Specifications
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:43.0 to 46.0 deg-C
- Specific rotation [a]20/D:+18.0 to +20.0 deg (C=3, Acetone)
- Solubility in Acetone:almost transparency
Specification Test Results
About this item
:
:230.26 g/mol
Boiling Pt:125°C
CAS Number:20881-04-3
Formula:C₁₁H₁₈O₅
MDL Number:MFCD00063224