Order Entry
Italy
ContactUsLinkComponent
Ethyl di-o-tolylphosphonoacetate ≥95.0% (by GC) Horner-Emmons reagent
Ethyl di-o-tolylphosphonoacetate ≥95.0% (by GC) Horner-Emmons reagent
Catalog # TCIAD2548-1G
Supplier:  TCI
CAS Number:  
Ethyl di-o-tolylphosphonoacetate ≥95.0% (by GC) Horner-Emmons reagent
Catalog # TCIAD2548-1G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Appearance:
    Colorless to Yellow to Orange clear liquid
  • Purity (GC):
    min. 95.0 %

Specification Test Results

About this item

 :
 :348.33 g/mol
Boiling Pt:240°C
CAS Number:188945-41-7
Formula:C₁₈H₂₁O₅P
MDL Number:MFCD01321166