
(R,R)-N,N'-Bis(trifluoromethanesulphonyl)-1,2-diphenylethylenediamine ≥98.0% (by GC)
Catalog # TCIAD2520-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:211.0 to 215.0 deg-C
- Specific rotation [a]20/D:+4.0 to +8.0 deg (C=1, CHCl3)
Specification Test Results
About this item
:
:476.41 g/mol
:GHS07
CAS Number:121788-73-6
Formula:C₁₆H₁₄F₆N₂O₄S₂
MDL Number:MFCD01321158