Order Entry
Italy
ContactUsLinkComponent
 
 
1,1-Diphenyl-2-propyn-1-ol ≥97.0% (by GC)
 :  TCI
 
 
1,1-Diphenyl-2-propyn-1-ol ≥97.0% (by GC)
 :  TCI

 

  • Appearance:
    White to Orange to Green powder to crystal
  • Purity (GC):
    min. 97.0 %
  • Melting point:
    47.0 to 51.0 deg-C
  • Solubility in Methanol:
    very faint turbidity

 

Frequently Bought Together