Order Entry
Italy
ContactUsLinkComponent
Diethyl dodecanedioate ≥97.0% (by GC)
Diethyl dodecanedioate ≥97.0% (by GC)
Catalog # TCIAD2486-25G
Supplier:  TCI
CAS Number:  
Diethyl dodecanedioate ≥97.0% (by GC)
Catalog # TCIAD2486-25G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    Colorless to Light yellow to Light orange clear liquid
  • Purity (GC):
    min. 97.0 %

Specification Test Results

About this item

 :
 :286.41 g/mol
EINECS:233-960-6
CAS Number:10471-28-0
Formula:C₁₆H₃₀O₄
MDL Number:MFCD00009204