1,3-Dibromoadamantane ≥97.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:876-53-9
:
Formula:C₁₀H₁₄Br₂
MDL Number:MFCD00077218
:294.03 g/mol
Specification Test Results
- Appearance:White to Orange to Green powder to crystal
- Purity (GC):min. 97.0 %
- Melting point:109.0 to 113.0 deg-C
- Solubility in hot Methanol:almost transparency
Specification Test Results
Frequently Bought Together