Order Entry
Italy
ContactUsLinkComponent
Dibutyltin bis(trifluoromethanesulphonate) ≥98.0% (W)
Corrosive
Toxic
Dibutyltin bis(trifluoromethanesulphonate) ≥98.0% (W)
Catalog # TCIAD2466-5G
Supplier:  TCI
CAS Number:  
Corrosive
Toxic
Dibutyltin bis(trifluoromethanesulphonate) ≥98.0% (W)
Catalog # TCIAD2466-5G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Appearance:
    White to Brown to Dark purple powder to crystal
  • Purity (Gravimetric Method):
    min. 98.0 %

Specification Test Results

About this item

 :
 :531.07 g/mol
UN:2923
ADR:8,III
 :GHS05
 :GHS06
CAS Number:38438-11-8
Formula:C₁₀H₁₈F₆O₆S₂Sn
MDL Number:MFCD00671529