Order Entry
Italy
ContactUsLinkComponent
2,3:5,6-Di-O-isopropylidene-D-mannofuranose ≥98.0% (by GC)
2,3:5,6-Di-O-isopropylidene-D-mannofuranose ≥98.0% (by GC)
Catalog # TCIAD2447-5G
Supplier:  TCI
CAS Number:  
2,3:5,6-Di-O-isopropylidene-D-mannofuranose ≥98.0% (by GC)
Catalog # TCIAD2447-5G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Melting point:
    122.0 to 126.0 deg-C
  • Specific rotation [a]20/D:
    +22.0 to +26.0 deg (C=1, Acetone)
  • Solubility in Methanol:
    almost transparency

Specification Test Results

About this item

 :
 :260.29 g/mol
CAS Number:14131-84-1
Formula:C₁₂H₂₀O₆
MDL Number:MFCD00134206