Order Entry
Italy
ContactUsLinkComponent
2,3-Dichloro-5,8-dihydroxy-1,4-naphthoquinone ≥97.0% (by HPLC, titration analysis)
Irritant
2,3-Dichloro-5,8-dihydroxy-1,4-naphthoquinone ≥97.0% (by HPLC, titration analysis)
Supplier:  TCI
Certificates
Irritant
2,3-Dichloro-5,8-dihydroxy-1,4-naphthoquinone ≥97.0% (by HPLC, titration analysis)
Supplier:  TCI

Specification Test Results

  • Appearance:
    Orange to Brown to Dark red powder to crystal
  • Purity (HPLC):
    min. 97.0 area%
  • Purity (Neutralization titration):
    min. 97.0 %
  • Melting point:
    197.0 to 201.0 deg-C

Specification Test Results

Frequently Bought Together