Order Entry
Italy
ContactUsLinkComponent
 
2-(1,3-Dioxolan-2-yl)furan ≥98.0% (by GC)
  TCIAD2417-25G
 :  TCI
 :  
2-(1,3-Dioxolan-2-yl)furan ≥98.0% (by GC)
  TCIAD2417-25G
 :  TCI
 :  

 

  • Pk:
    25 g
  • Seq:
    3308

 

 

  • Appearance:
    Colorless to Light orange to Yellow clear liquid
  • Purity (GC):
    min. 98.0 %

 

 

 :
 :140.14 g/mol
 :76°C
 :Refrigerator
 :1708-41-4
 :C₇H₈O₃
 :MFCD00671520