Trimethylene di(thiotosylate) ≥95.0% (by HPLC) protecting reagent for active methylene
Supplier: TCI
Certificates
About this item
CAS Number:3866-79-3
:
Formula:C₁₇H₂₀O₄S₄
MDL Number:MFCD00014918
:416.58 g/mol
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 95.0 area%
- Melting point:64.0 to 69.0 deg-C
Specification Test Results
Frequently Bought Together