Methyl-2,2-dimethoxypropionate ≥96.0% (by GC)
TCIAD2382-25G
: TCI
:
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 96.0 %
:
:148.1571 g/mol
:70°C
:233-211-3
:10076-48-9
:C₆H₁₂O₄
:MFCD00048066