2,2-Dimethoxypropionamide ≥98.0% (by total nitrogen basis)
Catalog # TCIAD2379-5G
Supplier: TCI
CAS Number:
Specifications
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (with Total Nitrogen):min. 98.0 %
- Melting point:115.0 to 118.0 deg-C
- Solubility in Methanol:almost transparency
Specification Test Results
About this item
:
:133.15 g/mol
Formula:C₅H₁₁NO₃
MDL Number:MFCD00671512