Order Entry
Italy
ContactUsLinkComponent
N,N'-Dibenzyl-4,13-diaza-18-crown 6-ether ≥98.0% (by GC, titration analysis)
Irritant
N,N'-Dibenzyl-4,13-diaza-18-crown 6-ether ≥98.0% (by GC, titration analysis)
Supplier:  TCI
Certificates
Irritant
N,N'-Dibenzyl-4,13-diaza-18-crown 6-ether ≥98.0% (by GC, titration analysis)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Purity (Nonaqueous Titration):
    min. 98.0 %
  • Melting point:
    82.0 to 85.0 deg-C

Specification Test Results

Related Information

Frequently Bought Together