Order Entry
Italy
ContactUsLinkComponent
5,6-Diacetoxyindole ≥98.0% (by HPLC, total nitrogen)
5,6-Diacetoxyindole ≥98.0% (by HPLC, total nitrogen)
Nome del fornitore:  TCI
Certificati
5,6-Diacetoxyindole ≥98.0% (by HPLC, total nitrogen)
Nome del fornitore:  TCI

Risultati dei test sulle specifiche

  • Appearance:
    White to Light yellow to Light red powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (with Total Nitrogen):
    min. 98.0 %
  • Melting point:
    136.0 to 140.0 deg-C
  • Solubility in Methanol:
    almost transparency

Risultati dei test sulle specifiche

Frequently Bought Together