Inserisci ordine
Italy
Orders LinkContactUsLinkComponent
3,3'-Dihydroxybenzidine ≥99.0% (by titrimetric analysis)

3,3'-Dihydroxybenzidine ≥99.0% (by titrimetric analysis)

Irritant
Health
Supplier:   TCI
3,3'-Dihydroxybenzidine ≥99.0% (by titrimetric analysis)
Supplier:  TCI
Certificates
3,3'-Dihydroxybenzidine ≥99.0% (by titrimetric analysis)

2 Options Available Below

Product Details & Documents
Synonyms:  4,4'-Diamino-3,3'-dihydroxybiphenyl, 3,3'-Dihydroxy-4,4'-diaminobiphenyl, 4,4'-Diamino-[1,1'-biphenyl]-3,3'-diol, Benzidine-3,3'-diol
  • CAS Number:2373-98-0
  • Formula:C₁₂H₁₂N₂O₂
  • MDL Number:MFCD00039149
  • Molecular Weight:216.24 g/mol

Documents

Specifications for Product Family

Specification Test Results

  • Appearance:
    White to Gray to Brown powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (Nonaqueous Titration):
    min. 99.0 %
  • Solubility in N,N-DMF:
    almost transparency
Product Family Options

Product Information

  • Conf
    Availability
    Price
  • 5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 77,00 €
    77,00 €
    1 * 5 g
       77,00 €

  • 25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 259,00 €
    259,00 €
    1 * 25 g
       259,00 €

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.