Order Entry
Italy
ContactUsLinkComponent
5,6-Diamino-2,3-dicyanopyrazine ≥98.0% (by total nitrogen basis)
Toxic
5,6-Diamino-2,3-dicyanopyrazine ≥98.0% (by total nitrogen basis)
Catalog # TCIAD2270-10G
Supplier:  TCI
CAS Number:  
Toxic
5,6-Diamino-2,3-dicyanopyrazine ≥98.0% (by total nitrogen basis)
Catalog # TCIAD2270-10G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    10 g
  • Seq:
    3236

Specifications

Specification Test Results

  • Appearance:
    Light orange to Yellow to Green powder to crystal
  • Purity (with Total Nitrogen):
    min. 98.0 %

Specification Test Results

About this item

 :
 :160.14 g/mol
Melting Pt:332°C
UN:2811
ADR:6.1,III
 :GHS06
CAS Number:36023-58-2
Formula:C₆H₄N₆
MDL Number:MFCD00191418