
2,4-Diamino-6-methoxy-1,3,5-triazine
Catalog # TCIAD2232-5G
Supplier: TCI
CAS Number:
Specifications
- Conf:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (Nonaqueous Titration):min. 98.0 %
- Solubility in hot Water:very faint turbidity
Specification Test Results
About this item
:
:141.13 g/mol
:GHS07
CAS Number:2827-45-4
Formula:C₄H₇N₅O
MDL Number:MFCD00191340