
2,5-Dimethylterephthalonitrile ≥99.0% (by GC)
Catalog # TCIAD2218-5G
Supplier: TCI
CAS Number:
Specifications
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 99.0 %
- Melting point:210.0 to 214.0 deg-C
Specification Test Results
About this item
:
:156.19 g/mol
:GHS07
CAS Number:39095-25-5
Formula:C₁₀H₈N₂
MDL Number:MFCD00191411