
3,4-Diethyl-2-ethoxycarbonyl-5-methylpyrrole ≥96.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:16200-50-3
:
Formula:C₁₂H₁₉NO₂
MDL Number:MFCD00075089
:209.29 g/mol
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 96.0 %
- Melting point:75.0 to 78.0 deg-C
- Solubility in Methanol:almost transparency
Specification Test Results
Frequently Bought Together