Order Entry
Italy
ContactUsLinkComponent
Di-p-tolylether ≥98.0% (by GC)
Irritant
Di-p-tolylether ≥98.0% (by GC)
Catalog # TCIAD2178-25G
Supplier:  TCI
CAS Number:  
Irritant
Di-p-tolylether ≥98.0% (by GC)
Catalog # TCIAD2178-25G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Melting point:
    47.0 to 51.0 deg-C
  • Solubility in Methanol:
    almost transparency

Specification Test Results

About this item

 :
 :198.27 g/mol
Boiling Pt:285°C
EINECS:216-423-0
 :GHS07
CAS Number:1579-40-4
Formula:C₁₄H₁₄O
MDL Number:MFCD00025979