
1,3-Diaminopyrene ≥98.0% (by HPLC)
Catalog # TCIAD2167-100MG
Supplier: TCI
CAS Number:
Specifications
- Pk:100 mg
- Seq:2440
Specifications
Specification Test Results
- Appearance:Yellow to Brown to Dark green powder to crystal
- Purity (HPLC):min. 98.0 area%
- Elemental analysis (Nitrogen):11.50 to 12.50 %
Specification Test Results
About this item
:
:232.29 g/mol
:GHS07
CAS Number:92821-64-2
Formula:C₁₆H₁₂N₂
MDL Number:MFCD00142518