
1,1'-Diphenyl-4,4'-bipyridinium dichloride ≥97.0% (by HPLC, titration analysis)
Catalog # TCIAD2165-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:White to Gray to Brown powder to crystal
- Purity (HPLC):min. 97.0 area%
- Purity (Nonaqueous Titration):min. 97.0 %
Specification Test Results
About this item
:
:381.30 g/mol
:GHS07
CAS Number:47369-00-6
Formula:C₂₂H₁₈Cl₂N₂
MDL Number:MFCD00142355