
2,2'-Dihydroxydiphenyl ether ≥98.0% (by GC)
Catalog # TCIAD2150-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:121.0 to 125.0 deg-C
- Solubility in Methanol:very faint turbidity
Specification Test Results
About this item
:
:202.21 g/mol
:GHS07
CAS Number:15764-52-0
Formula:C₁₂H₁₀O₃
MDL Number:MFCD00142736