Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Italy
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
S,S'-Diethyl dithiocarbonate
S,S'-Diethyl dithiocarbonate ≥98.0% (by GC)
S,S'-Diethyl dithiocarbonate ≥98.0% (by GC)
TCIAD2078-10G
:
TCI
:
S,S'-Diethyl dithiocarbonate ≥98.0% (by GC)
TCIAD2078-10G
:
TCI
:
Pk:
10 g
Seq:
3236
Appearance:
Colorless to Light yellow clear liquid
Purity (GC):
min. 98.0 %
:
:
150.25 g/mol
:
168°C
:
623-80-3
:
C₅H₁₀OS₂
:
MFCD00144162