
3,5-Di(2-pyridyl)pyrazole ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:129485-83-2
:
Formula:C₁₃H₁₀N₄
MDL Number:MFCD00130125
:222.25 g/mol
:GHS07
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Nonaqueous Titration):min. 98.0 %
- Melting point:188.0 to 192.0 deg-C
- Solubility in hot Methanol:within almost transparency
Specification Test Results
Frequently Bought Together