
2,5-Dimethyl-7,7,8,8-tetracyanoquinodimethane ≥98.0% (by total nitrogen basis)
: TCI
:1487-82-7
:
:C₁₄H₈N₄
:MFCD00130123
:6.1,III
:232.25 g/mol
:3439
:GHS06
:Refrigerator
- Appearance:Light yellow to Brown powder to crystal
- Purity (with Total Nitrogen):min. 98.0 %
- Solubility in hot Acetonitrile:almost transparency
Frequently Bought Together