Order Entry
Italy
ContactUsLinkComponent
1,3-Diiminobenz[f]isoindoline ≥95.0% (by total nitrogen basis)
Irritant
1,3-Diiminobenz[f]isoindoline ≥95.0% (by total nitrogen basis)
Catalog # TCIAD2009-1G
Supplier:  TCI
CAS Number:  
Irritant
1,3-Diiminobenz[f]isoindoline ≥95.0% (by total nitrogen basis)
Catalog # TCIAD2009-1G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Appearance:
    Orange to Green to Amber powder to crystal
  • Purity (with Total Nitrogen):
    min. 95.0 %

Specification Test Results

About this item

 :
 :195.23 g/mol
 :GHS07
CAS Number:65558-69-2
Formula:C₁₂H₉N₃
MDL Number:MFCD00059067