Order Entry
Italy
ContactUsLinkComponent
Diallyldimethylammonium chloride 60% in water
Diallyldimethylammonium chloride 60% in water
Supplier:  TCI
Certificates
undefined
Diallyldimethylammonium chloride 60% in water
Supplier:  TCI

Specification Test Results

  • Appearance:
    Colorless to Light yellow to Light orange clear liquid
  • Concentration (Bromination Method):
    60.0 to 66.0 w/w%

Specification Test Results

Related Information

Frequently Bought Together