
4,5-Dicarboxy-γ-pentadecanolactone ≥98.0% (by titrimetric analysis)
Catalog # TCIAD1963-25G
Supplier: TCI
CAS Number:
Specifications
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (Neutralization titration):min. 98.0 %
- Melting point:144.0 to 148.0 deg-C
- Specific rotation [a]20/D:-11.0 to -14.0 deg (C=3, EtOH)
- Solubility in Methanol:almost transparency
Specification Test Results
About this item
:
:328.41 g/mol
:GHS07
CAS Number:469-77-2
Formula:C₁₇H₂₈O₆
MDL Number:MFCD00059059