1,2:5,6-Di-O-isopropylidene-α-D-glucofuranose ≥97.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:582-52-5
:
Formula:C₁₂H₂₀O₆
MDL Number:MFCD00005544
:260.29 g/mol
Merck Index:14,02965
EINECS:209-486-0
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 97.0 %
- Melting point:108.0 to 112.0 deg-C
- Specific rotation [a]20/D:-17.0 to -20.0 deg (C=5, H2O)
- Solubility in hot Water:very faint turbidity
Specification Test Results
Related Information
Frequently Bought Together