
2,5-Dimethyltetrahydrofuran ≥98.0% (by GC) stabilised
TCIAD1948-25ML
: TCI
:
- Pack type:Glass bottle
- Pk:25 mL
- Seq:2315
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 98.0% (total of isomer)
:
:92°C
:27°C
:1993
:3,III
:Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol)
:213-707-6
:GHS02
:1003-38-9
:C₆H₁₂O₃
:MFCD00005369