2,4-Dimethyl-2,4-pentanediol ≥96.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:24892-49-7
:
Formula:C₇H₁₆O₂
MDL Number:MFCD00012306
:132.20 g/mol
Boiling Pt:100°C
Flash Pt:85°C
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:Colorless to Light yellow clear liquid
- Purity (GC):min. 96.0 %
Specification Test Results
Frequently Bought Together