Order Entry
Italy
ContactUsLinkComponent
9,10-Dihydro-9-oxa-10-phosphaphenanthrene-10-oxide ≥97.0% (by titrimetric analysis)
Irritant
9,10-Dihydro-9-oxa-10-phosphaphenanthrene-10-oxide ≥97.0% (by titrimetric analysis)
Supplier:  TCI
Certificates
Irritant
9,10-Dihydro-9-oxa-10-phosphaphenanthrene-10-oxide ≥97.0% (by titrimetric analysis)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (Neutralization titration):
    min. 97.0 %
  • Melting point:
    116.0 to 120.0 deg-C

Specification Test Results

Frequently Bought Together