

1,3-Divinyltetramethyldisiloxane ≥98.0% (by GC)
: TCI
:2627-95-4
:
:C₈H₁₈OSi₂
:MFCD00014933
:186.40 g/mol
:139°C
:24°C
:220-099-6
:1993
:3,III
:GHS02
:GHS07
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 98.0 %
Frequently Bought Together