
4,4'-Dithiodibutyric acid ≥95.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:2906-60-7
:
Formula:C₈H₁₄O₄S₂
MDL Number:MFCD00004406
:238.32 g/mol
EINECS:220-818-3
:GHS07
Specification Test Results
- Appearance:White to Light yellow to Dark green powder to crystal
- Purity (GC):min. 95.0 %
- Purity (Neutralization titration):min. 95.0 %
- Melting point:107.0 to 111.0 deg-C
Specification Test Results
Frequently Bought Together