
2,5-Dichloroterephthalonitrile ≥99.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:1897-43-4
:
Formula:C₈H₂Cl₂N₂
MDL Number:MFCD00059584
:197.02 g/mol
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 99.0 %
- Melting point:210.0 to 217.0 deg-C
Specification Test Results
Frequently Bought Together