

N,N,2,2-Tetramethyl-1,3-propanediamine ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:53369-71-4
:
Formula:C₇H₁₈N₂
MDL Number:MFCD00044018
:130.24 g/mol
Boiling Pt:153°C
Flash Pt:35°C
EINECS:258-501-7
UN:2734
ADR:8,II
:GHS02
:GHS05
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
Specification Test Results
Frequently Bought Together