
Diethylene glycol dibenzoate ≥97.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:120-55-8
:
Formula:C₁₈H₁₈O₅
MDL Number:MFCD00020679
:314.34 g/mol
Flash Pt:228°C
EINECS:204-407-6
:GHS07
Specification Test Results
- Appearance:Colorless to Light yellow to Light orange clear liquid
- Purity (GC):min. 97.0 %
Specification Test Results
Frequently Bought Together