


Dodecene-1 LAS ≥98.0% (by titrimetric analysis)
TCIAD1428-1G
: TCI
:
- Pk:1 g
- Seq:3047
- Appearance:White to Light yellow powder to crystal
- Purity (Ion exchange titration):min. 98.0 %
:
:348.48 g/mol
:14,08612
:246-680-4
:3077
:9,III
:GHS05
:GHS07
:GHS09
:68628-60-4
:C₁₈H₂₉NaO₃S