
1,2-Diaminopropane-N,N,N',N'-tetraacetic acid ≥98.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:4408-81-5
:
Formula:C₁₁H₁₈N₂O₈
MDL Number:MFCD00004286
:306.27 g/mol
EINECS:224-559-7
:GHS07
Specification Test Results
- Appearance:White powder to crystal
- Purity (Chelometric Titration):min. 98.0 %
- Solubility in Sodium hydroxide solution:almost transparency
Specification Test Results
Frequently Bought Together